Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015135.2 | NP_055950.1 | 41 | 5764 | ||
XM_005250235.3 | XP_005250292.1 | 40 | 5716 | |||
GenBank mRNA | AK293497.1 | - | 19 | 2551 | ||
BC044255.1 | - | 16 | 2194 | |||
BC063612.1 | - | 13 | 1548 | |||
BC073947.1 | - | 9 | 1002 | |||
BC094868.1 | - | 7 | 702 | |||
BC136624.1 | - | 41 | 5759 | 9020614 | ||
BC146784.1 | - | 41 | 5738 | 9007240 | ||
D86978.1 | - | 41 | 5738 |