Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144868.3 | NP_659117.2 | 7 | 2506 | ||
GenBank mRNA | AF237953.1 | - | 5 | 523 | ||
AK171516.1 | - | 6 | 2745 | |||
BC025838.1 | - | 1 | 39 | |||
BC038056.1 | - | 2 | 614 | |||
BC054751.1 | - | 7 | 1785 | |||
BC055289.1 | - | 8 | 2309 | |||
BC096538.1 | - | 7 | 2506 |