Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: METAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015143.2 | NP_055958.2 | 6, 7 | 630 | ||
XM_011531777.2 | XP_011530079.1 | 6, 7 | 627 | |||
XM_011531778.1 | XP_011530080.1 | 6, 7 | 423 | |||
XM_011531779.1 | XP_011530081.1 | 6, 7 | 630 | |||
XM_017007914.1 | XP_016863403.1 | 6, 7 | 630 | |||
XM_017007915.1 | XP_016863404.1 | 7, 8 | 644 | |||
XM_017007916.1 | XP_016863405.1 | 6, 7 | 643 | |||
XM_017007917.1 | XP_016863406.1 | 6, 7 | 419 | |||
GenBank mRNA | AK304239.1 | - | 6, 7 | 546 | ||
BC030054.1 | - | 2, 3 | 188 | 3958447 | ||
CR936632.1 | - | 8, 9 | 4208 | |||
D42084.1 | - | 6, 7 | 538 |