Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A530064D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113556.1 | NP_001107028.1 | 2 | 597 | ||
NM_178796.6 | NP_848911.2 | 2 | 597 | |||
GenBank mRNA | AB372005.1 | - | 2 | 374 | ||
AK080114.1 | - | 2 | 558 | |||
AK089248.1 | - | 2 | 611 | |||
AK089332.1 | - | 2 | 558 | |||
AK170607.1 | - | 2 | 616 | |||
AK171433.1 | - | 558 | ||||
BC139185.1 | - | 2 | 432 | 8862207 | ||
BC139186.1 | - | 2 | 432 | 8862208 | ||
BC145572.1 | - | 2 | 432 | 9054138 | ||
EU003111.1 | - | 2 | 328 | |||
EU003112.1 | - | 2 | 328 |