Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113356.1 | NP_001106827.1 | 1257 | |||
GenBank mRNA | AF459018.1 | - | 1257 |
Gene Symbol: C1ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023143.3 | NP_075632.3 | 9 | 1408 | ||
GenBank mRNA | AF148216.1 | - | 9 | 1396 | ||
BC004637.1 | - | 9 | 1350 | 3586055 | ||
BC093528.1 | - | 9 | 1624 | 5135606 | ||
BC172078.1 | - | 9 | 1333 | 9054364 |