Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fbxo38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134136.3 | NP_598897.2 | 12, 13 | 1793 | ||
GenBank mRNA | AK031096.1 | - | 12, 13 | 1743 | ||
AK031347.1 | - | 12, 13 | 1724 | |||
AK034761.1 | - | 7, 8 | 977 | |||
AK089046.1 | - | 12, 13 | 1765 | |||
AY267463.1 | - | 12, 13 | 1726 | |||
BC056348.1 | - | 12, 13 | 1745 | 5721407 |