Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009802.2 | NP_033932.2 | 4, 5 | 532 | ||
GenBank mRNA | AF079834.1 | - | 3, 4 | 622 | ||
AF079835.1 | - | 4, 5 | 559 | |||
AK132599.1 | - | 4, 5 | 533 | |||
AK161216.1 | - | 4, 5 | 528 | |||
BC006726.1 | - | 4, 5 | 619 | |||
BC018582.1 | - | 4, 5 | 509 | |||
BC036270.1 | - | 4, 5 | 515 | |||
BC039657.1 | - | 4, 5 | 517 | |||
BC046495.1 | - | 3, 4 | 308 | |||
BC049973.1 | - | 4, 5 | 530 | |||
BC055437.1 | - | 4, 5 | 518 | 4190566 |