Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1b26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010644.2 | NP_034774.1 | 4 | 601 | ||
GenBank mRNA | AK075752.1 | - | 2 | 157 | ||
AK075766.1 | - | 2 | 157 | |||
BC024688.1 | - | 4 | 575 | 4167696 | ||
BC138704.1 | - | 4 | 583 | 8861726 | ||
K01831.1 | - | 4 | 601 | |||
V00828.1 | - | 225 | ||||
X63327.1 | - | 4 | 580 |
Gene Symbol: Egfbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010115.6 | NP_034245.3 | 4 | 583 | ||
GenBank mRNA | AB016032.1 | - | 4 | 559 | ||
X58628.1 | - | 4 | 583 |