Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168289.1 | NP_001161761.1 | 3 | 767 | ||
NM_027772.2 | NP_082048.2 | 3 | 767 | |||
GenBank mRNA | AB210840.1 | - | 3 | 491 | ||
AK017329.1 | - | 3 | 737 | |||
AK140091.1 | - | 3 | 654 | |||
AK165732.1 | - | 3 | 766 | |||
BC147693.1 | - | 3 | 544 | 9056666 | ||
BC147694.1 | - | 3 | 544 | 9056667 | ||
BC147752.1 | - | 3 | 517 | 9056701 | ||
BC147753.1 | - | 3 | 517 | 9056702 | ||
BC172142.1 | - | 3 | 470 |