Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024215.2 | NP_077177.2 | 2 | 337 | ||
GenBank mRNA | AK005247.1 | - | 2 | 339 | ||
AK014081.1 | - | 2 | 287 | |||
AK133585.1 | - | 2 | 335 | |||
AK156931.1 | - | 2 | 272 | |||
BC003465.1 | - | 2 | 274 | 2648747 |