Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdpoz5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207273.2 | NP_997156.2 | 213 | |||
GenBank mRNA | BC111104.1 | - | 1 | 211 |
Gene Symbol: Gm10697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163731.1 | NP_001157203.1 | 1 | 213 |