Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sclt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081411.1 | NP_001074880.1 | 18 | 2282 | ||
GenBank mRNA | AK083894.1 | - | 16 | 2338 | ||
AK089007.1 | - | 7 | 774 | |||
BC043136.1 | - | 18 | 2266 | 6812105 | ||
BC060271.1 | - | 18 | 2266 | 6812105 |