Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005813.4 | NP_005804.1 | 10 | 1974 | ||
XM_005264237.3 | XP_005264294.1 | 11 | 2247 | |||
XM_005264238.3 | XP_005264295.1 | 10 | 1223 | |||
XM_017003741.1 | XP_016859230.1 | 12 | 2252 | |||
XM_017003742.1 | XP_016859231.1 | 11 | 1291 | |||
XM_017003743.1 | XP_016859232.1 | 8 | 973 | |||
XM_017003744.1 | XP_016859233.1 | 7 | 721 | |||
GenBank mRNA | AB015982.2 | - | 10 | 1974 | ||
AK027585.1 | - | 7 | 783 | |||
AK122883.1 | - | 3 | 1648 | |||
BC030706.2 | - | 10 | 1919 | 5266769 |