Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139222.3 | NP_631968.1 | 2 | 281 | ||
GenBank mRNA | AJ437648.1 | - | 2 | 281 |