Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081169.1 | NP_001074638.1 | 7 | 861 | ||
GenBank mRNA | BC089617.1 | - | 7 | 688 | ||
BC126962.1 | - | 7 | 679 | 40092851 |