Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist2h3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178216.3 | NP_835734.2 | 1 | 436 | ||
GenBank mRNA | BC094041.1 | - | 436 | 5099464 |
Gene Symbol: Hist2h3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178215.2 | NP_835587.1 | 1 | 275 | ||
GenBank mRNA | BC139251.1 | - | 1 | 275 | 8862273 | |
BC139252.1 | - | 1 | 275 | 8862274 |
Gene Symbol: Hist2h3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054045.4 | NP_473386.1 | 1 | 421 | ||
GenBank mRNA | BC015270.1 | - | 1 | 421 | 4240748 | |
BC132488.1 | - | 1 | 265 | 40130765 | ||
BC132490.1 | - | 1 | 265 | 40130767 |