Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEL1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297592.1 | NP_001284521.1 | 19 | 2757 | ||
NM_001297594.1 | NP_001284523.1 | 19 | 2779 | |||
NM_015187.4 | NP_056002.2 | 19 | 2974 | |||
XM_011513819.2 | XP_011512121.2 | 19 | 2968 | |||
GenBank mRNA | AB018289.1 | - | 18 | 2472 | ||
AK025282.1 | - | 7 | 640 | |||
AK289687.1 | - | 19 | 2722 | |||
AK300219.1 | - | 10 | 1072 | |||
BC009945.2 | - | 12 | 1489 | |||
BC041046.1 | - | 10 | 1126 | |||
BC060867.1 | - | 19 | 2767 | 30344157 | ||
CR749817.1 | - | 19 | 2757 |