Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013833.3 | NP_001013855.1 | 4 | 796 | ||
NM_011160.3 | NP_035290.1 | 4 | 719 | |||
GenBank mRNA | AF084547.1 | - | 4 | 719 | ||
AK083991.1 | - | 4 | 796 | |||
BC113162.1 | - | 4 | 701 | 40060049 |