Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TBC1D12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015188.1 | NP_056003.1 | 9 | 1895 | ||
XM_006717732.3 | XP_006717795.2 | 8 | 1814 | |||
XM_006717733.2 | XP_006717796.1 | 10 | 855 | |||
XM_011539558.2 | XP_011537860.1 | 9 | 906 | |||
XM_017015996.1 | XP_016871485.1 | 7 | 1670 | |||
XM_017015998.1 | XP_016871487.1 | 8 | 682 | |||
XM_017015999.1 | XP_016871488.1 | 8 | 898 | |||
GenBank mRNA | AB011180.1 | - | 9 | 1711 | ||
AB449898.1 | - | 9 | 1692 | |||
AK090865.1 | - | 2 | 1762 | |||
AL049956.1 | - | 8 | 643 |