Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Khdc1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033904.1 | NP_001029076.1 | 3 | 719 | ||
GenBank mRNA | BC099381.1 | - | 3 | 719 | 30937206 | |
BC141037.1 | - | 413 | 9055649 |
Gene Symbol: Khdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183322.2 | NP_899145.2 | 3 | 735 | ||
GenBank mRNA | AK136166.1 | - | 3 | 662 | ||
AY238603.1 | - | 3 | 498 |