Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm2002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100596.1 | NP_001094066.1 | 14 | 2298 |
Gene Symbol: Il11ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010550.3 | NP_034680.3 | 14 | 2298 |