Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013838.2 | NP_038866.2 | 6 | 1810 | ||
GenBank mRNA | AF057748.1 | - | 6 | 1379 | ||
AK137447.1 | - | 5 | 1038 | |||
BC141131.1 | - | 6 | 1681 | 9055743 | ||
U49069.1 | - | 6 | 1810 |