Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025330.3 | NP_079606.3 | 9 | 1336 | ||
GenBank mRNA | AK002831.1 | - | 9 | 1289 | ||
AK010556.1 | - | 5 | 1001 | |||
BC064042.1 | - | 5 | 1119 | 1332242 | ||
BC099530.1 | - | 5 | 1005 |