Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Whsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081102.2 | NP_001074571.2 | 13 | 2694 | ||
NM_175231.2 | NP_780440.2 | 13 | 2691 | |||
GenBank mRNA | AK078622.1 | - | 10 | 1330 | ||
AK129287.1 | - | 10 | 1629 | |||
BC046473.1 | - | 8 | 1120 | |||
BC053454.1 | - | 2 | 243 | |||
BC079668.1 | - | 13 | 2717 |