Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111096.1 | NP_001104566.1 | 6 | 722 | ||
NM_010747.2 | NP_034877.2 | 6 | 659 | |||
GenBank mRNA | AK028112.1 | - | 6 | 655 | ||
AK151914.1 | - | 6 | 720 | |||
AK152199.1 | - | 6 | 720 | |||
AK153038.1 | - | 6 | 656 | |||
AK170561.1 | - | 6 | 720 | |||
BC003253.1 | - | 6 | 599 | 3500419 | ||
BC031547.1 | - | 6 | 692 | 3589207 | ||
BC157998.1 | - | 6 | 491 | 9007404 | ||
M64608.1 | - | 6 | 708 |