Gene Symbol: Flnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081185.1 | NP_001074654.1 | 30 | 5416 | ||
GenBank mRNA | AF353670.1 | - | 1 | 52 | ||
BC052186.1 | - | 3 | 365 | |||
BC060276.1 | - | 10 | 1576 | |||
BC151097.1 | - | 30 | 5221 | 9088010 | ||
BC158128.1 | - | 30 | 5221 | 9007396 | ||
BC172159.1 | - | 30 | 5221 |