Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015202.3 | NP_056017.3 | 5 | 352 | ||
XM_005255201.3 | XP_005255258.1 | 5 | 371 | |||
XM_005255202.3 | XP_005255259.1 | 5 | 371 | |||
XM_005255203.3 | XP_005255260.1 | 5 | 371 | |||
XM_005255206.3 | XP_005255263.1 | 5 | 371 | |||
XM_006721025.3 | XP_006721088.1 | 5 | 371 | |||
XM_011545773.2 | XP_011544075.1 | 5 | 341 | |||
XM_011545774.1 | XP_011544076.1 | 4 | 287 | |||
XM_011545775.2 | XP_011544077.1 | 5 | 371 | |||
XM_011545776.2 | XP_011544078.1 | 5 | 371 | |||
XM_017023085.1 | XP_016878574.1 | 5 | 371 | |||
XM_017023086.1 | XP_016878575.1 | 4 | 305 | |||
XM_017023087.1 | XP_016878576.1 | 5 | 371 | |||
XM_017023088.1 | XP_016878577.1 | 3 | 265 | |||
GenBank mRNA | AB011128.2 | - | 5 | 350 | ||
BC150276.1 | - | 5 | 350 | 9007250 | ||
BX472933.1 | - | 5 | 364 |