Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 9830147E19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242388.1 | NP_001229317.1 | 4 | 2580 | ||
GenBank mRNA | AK141688.1 | - | 5 | 2431 | ||
AK162659.1 | - | 3 | 2436 |
Gene Symbol: Gm2381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK049289.1 | - | 4 | 1978 |