Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdfy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111279.1 | NP_001104749.1 | 3, 4 | 356 | ||
NM_027057.3 | NP_081333.1 | 3, 4 | 356 | |||
GenBank mRNA | AK005937.1 | - | 3, 4 | 320 | ||
AK007215.1 | - | 3, 4 | 357 | |||
AK157204.1 | - | 3, 4 | 315 | |||
AK159710.1 | - | 3, 4 | 327 | |||
AK165554.1 | - | 3, 4 | 328 | |||
AK220391.1 | - | 3, 4 | 290 | |||
BC025226.1 | - | 3, 4 | 270 | 5011404 |