Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdfy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111279.1 | NP_001104749.1 | 4 | 388 | ||
NM_027057.3 | NP_081333.1 | 4 | 388 | |||
GenBank mRNA | AK005937.1 | - | 4 | 352 | ||
AK007215.1 | - | 4 | 389 | |||
AK157204.1 | - | 4 | 347 | |||
AK159710.1 | - | 4 | 359 | |||
AK165554.1 | - | 4 | 360 | |||
AK190401.1 | - | 1 | 32 | |||
AK220391.1 | - | 4 | 322 | |||
BC025226.1 | - | 4 | 302 | 5011404 |