Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009942.2 | NP_034072.2 | 3 | 293 | ||
GenBank mRNA | AK008339.1 | - | 3 | 214 | ||
AK131673.1 | - | 3 | 214 | |||
AK135174.1 | - | 3 | 222 | |||
BC005564.1 | - | 233 | ||||
BC096048.1 | - | 3 | 208 | 3465479 | ||
BC099474.1 | - | 3 | 222 | 30933585 | ||
CJ075429.1 | - | 3 | 298 | |||
X53157.1 | - | 3 | 202 |
Gene Symbol: Gm11273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC023228.1 | - | 221 |