Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mup13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134674.1 | NP_001128146.1 | 2 | 285 |
Gene Symbol: Mup17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200006.1 | NP_001186935.1 | 2 | 231 |