Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nat14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201355.3 | NP_958743.1 | 3 | 1400 | ||
GenBank mRNA | AK158627.1 | - | 3 | 1401 | ||
AK163549.1 | - | 3 | 1130 | |||
BC047219.1 | - | 3 | 1036 | 6491365 |