Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-T22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010397.4 | NP_034527.1 | 7 | 1306 | ||
GenBank mRNA | AB359229.1 | - | 7 | 1051 | ||
AK133985.1 | - | 8 | 1238 | |||
AK162874.1 | - | 7 | 1103 | |||
BC003819.1 | - | 7 | 1080 | 3484613 | ||
M35243.1 | - | 7 | 1275 | |||
M35245.1 | - | 7 | 1306 | |||
M35247.1 | - | 7 | 1268 |
Gene Symbol: H2-T10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010395.7 | NP_034525.4 | 1201 | |||
GenBank mRNA | AK088241.1 | - | 6 | 703 | ||
AK153876.1 | - | 1190 | ||||
AK161731.1 | - | 6 | 1517 | |||
BC021751.1 | - | 1295 | 4165944 | |||
BC021760.1 | - | 1319 | 5055493 | |||
M35244.1 | - | 7 | 1305 | |||
M35246.1 | - | 8 | 1193 |
Gene Symbol: H2-T9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010399.2 | NP_034529.1 | 1187 | |||
GenBank mRNA | BC044208.1 | - | 1198 | 5036676 | ||
M35248.1 | - | 1187 |