Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198654.3 | NP_941056.3 | 6 | 1632 | ||
GenBank mRNA | AK167096.1 | - | 169 | |||
BC029086.1 | - | 6 | 1616 | 3156934 |