Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MCF2L SIRNA Assays ›
Gene Symbol:
MCF2L
Gene Name:
MCF.2 cell line derived transforming sequence like
Gene Aliases:
ARHGEF14, DBS, OST
Chromosome Location:
Chr.13: 112894378 - 113099739 on Build GRCh38
UniGene ID:
Hs.170422
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23426
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MCF2L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001112732.2
NP_001106203.2
6
675
NM_001320815.1
NP_001307744.1
6
707
NM_001320816.1
NP_001307745.1
6
884
NM_001320817.1
NP_001307746.1
6
707
NM_024979.4
NP_079255.4
6
707
XM_011537482.1
XP_011535784.1
7
664
XM_011537483.2
XP_011535785.1
7
784
XM_011537484.1
XP_011535786.1
6
1111
XM_011537485.1
XP_011535787.1
6
783
XM_011537486.2
XP_011535788.2
6
799
XM_011537487.1
XP_011535789.1
6
704
XM_011537488.1
XP_011535790.1
6
543
XM_011537490.1
XP_011535792.1
6
1077
XM_011537491.1
XP_011535793.1
4
492
XM_017020493.1
XP_016875982.1
6
799
XM_017020494.1
XP_016875983.1
6
799
XM_017020495.1
XP_016875984.1
6
799
XM_017020496.1
XP_016875985.1
4
492
XM_017020497.1
XP_016875986.1
6
1077
XM_017020498.1
XP_016875987.1
4
492
XM_017020499.1
XP_016875988.1
6
799
GenBank mRNA
AB002360.1
-
6
652
AB116074.1
-
6
563
AB116075.1
-
1
39
AK094768.1
-
4
472
AK295047.1
-
6
704
AK295670.1
-
6
517
AK307867.1
-
6
855
AK315969.1
-
6
675
BC011853.2
-
6
667
4138009
BC020208.1
-
6
667
4121441
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of Rho protein signal transduction
intracellular signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
phosphatidylinositol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0