Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmigd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174169.2 | NP_001167640.1 | 2 | 766 | ||
GenBank mRNA | BC100416.1 | - | 2 | 766 | 30916707 |
Gene Symbol: Adora3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009631.4 | NP_033761.2 | 2 | 1344 | ||
GenBank mRNA | AK013534.1 | - | 2 | 1344 | ||
AK154312.1 | - | 2 | 1346 |
Set Membership: |
![]() |