Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mup10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122647.1 | NP_001116119.1 | 2 | 281 | ||
GenBank mRNA | AJ309921.1 | - | 214 | |||
CB948918.1 | - | 190 | ||||
EU882231.1 | - | 2 | 214 |
Gene Symbol: LOC100048884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC055353.1 | - | 261 |
Gene Symbol: Mup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC099597.1 | - | 262 | 947916 | ||
X04115.1 | - | 83 |
Gene Symbol: Mup2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC012259.1 | - | 273 | 4196403 | ||
BC059097.1 | - | 259 | 4159984 | |||
M16356.1 | - | 272 |