Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039696.1 | NP_001034785.1 | 4 | 872 | ||
GenBank mRNA | AJ972670.1 | - | 4 | 872 |
Gene Symbol: Rhox4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039688.3 | NP_001034777.1 | 4 | 834 | ||
GenBank mRNA | AJ972665.1 | - | 4 | 834 | ||
AK017380.1 | - | 4 | 749 | |||
BC057925.1 | - | 4 | 749 | 5292192 | ||
DQ058641.1 | - | 4 | 809 |
Gene Symbol: Rhox4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021300.2 | NP_067275.1 | 4 | 829 | ||
GenBank mRNA | AF265350.1 | - | 4 | 817 | ||
AJ972666.1 | - | 4 | 829 |
Gene Symbol: Rhox4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039695.1 | NP_001034784.1 | 702 | |||
GenBank mRNA | AJ972668.1 | - | 702 |
Gene Symbol: Rhox4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201236.3 | NP_957688.2 | 4 | 920 | ||
GenBank mRNA | AJ972669.1 | - | 4 | 920 | ||
BC094891.1 | - | 4 | 882 | 30019143 |
Gene Symbol: Rhox4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039698.1 | NP_001034787.1 | 4 | 921 | ||
GenBank mRNA | AJ972671.1 | - | 4 | 921 |