Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028677.4 | NP_082953.1 | 10 | 733 | ||
GenBank mRNA | AK003179.1 | - | 10 | 711 | ||
AK005202.1 | - | 10 | 713 | |||
BC050116.1 | - | 10 | 687 | 6432881 |