Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rplp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018853.3 | NP_061341.1 | 3 | 420 | ||
GenBank mRNA | AK007832.1 | - | 3 | 451 | ||
AK021242.1 | - | 11 | ||||
AK169347.1 | - | 3 | 450 | |||
BC058685.1 | - | 3 | 462 | 6410847 | ||
BC092536.1 | - | 420 | 6774435 |