Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC115446.1 | - | 1 | 78 |
Gene Symbol: Hist1h4m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195421.1 | NP_001182350.1 | 1 | 123 |
Gene Symbol: Hist1h4n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175657.2 | NP_783588.1 | 1 | 85 | ||
GenBank mRNA | BC115448.2 | - | 1 | 85 | 40099764 | |
BC152397.1 | - | 1 | 85 | 40099768 |