Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bglap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037939.2 | NP_001033028.1 | 3 | 261 | ||
GenBank mRNA | BY351185.1 | - | 3 | 260 | ||
CJ326768.1 | - | 1 | 59 |