Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Galnt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107151.2 | NP_001100621.1 | 6 | 716 | ||
NM_001122644.1 | NP_001116116.1 | 11 | 2028 | |||
XM_008769333.1 | XP_008767555.1 | 11 | 2005 | |||
XM_008769334.1 | XP_008767556.1 | 11 | 2027 |