Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012542.2 | NP_036674.1 | 5 | 754 | ||
GenBank mRNA | J02852.1 | - | 5 | 733 |