Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017150.1 | NP_058846.1 | 4 | 561 | ||
GenBank mRNA | BC086404.1 | - | 4 | 555 | 7304319 | |
FQ221016.1 | - | 4 | 562 | |||
FQ221647.1 | - | 4 | 568 | |||
FQ229056.1 | - | 4 | 566 | |||
FQ229057.1 | - | 4 | 566 | |||
X68283.1 | - | 4 | 561 |