Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk2ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113751.1 | NP_001107223.1 | 3, 4 | 505 | ||
XM_008769238.1 | XP_008767460.1 | 3, 4 | 835 | |||
XM_008769239.1 | XP_008767461.1 | 3, 4 | 759 | |||
GenBank mRNA | BC158683.1 | - | 3, 4 | 505 | 8373627 |