Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135085.1 | NP_001128557.1 | 4 | 347 | ||
GenBank mRNA | BC168750.1 | - | 4 | 313 | 6921773 | |
FQ217038.1 | - | 4 | 313 | |||
FQ221105.1 | - | 4 | 345 | |||
FQ221517.1 | - | 4 | 313 | |||
FQ221580.1 | - | 4 | 374 | |||
FQ221827.1 | - | 4 | 315 | |||
FQ222397.1 | - | 4 | 313 | |||
FQ222427.1 | - | 4 | 316 | |||
FQ222653.1 | - | 4 | 362 | |||
FQ223239.1 | - | 4 | 313 | |||
FQ223409.1 | - | 4 | 362 | |||
FQ228483.1 | - | 4 | 316 | |||
FQ228634.1 | - | 4 | 321 | |||
FQ228635.1 | - | 4 | 358 |
Gene Symbol: LOC687679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748839.2 | XP_003748887.1 | 1 | 314 |