Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox6a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012812.3 | NP_036944.2 | 3 | 452 | ||
XM_006230218.1 | XP_006230280.1 | 4 | 501 | |||
GenBank mRNA | FM058713.1 | - | 4 | 506 | ||
FM121690.1 | - | 3 | 358 | |||
FQ217951.1 | - | 3 | 402 | |||
FQ217987.1 | - | 3 | 359 | |||
FQ223608.1 | - | 3 | 408 | |||
FQ224015.1 | - | 3 | 410 | |||
FQ224196.1 | - | 3 | 409 | |||
FQ224467.1 | - | 3 | 398 | |||
FQ224572.1 | - | 3 | 417 | |||
X12554.1 | - | 3 | 372 |