Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021265.3 | NP_067088.2 | 17 | 1339 | ||
XM_006245011.2 | XP_006245073.1 | 18 | 2626 | |||
XM_008767130.1 | XP_008765352.1 | 17 | 2605 | |||
XM_008767131.1 | XP_008765353.1 | 19 | 1766 | |||
XM_008767132.1 | XP_008765354.1 | 20 | 1652 | |||
XM_008767133.1 | XP_008765355.1 | 16 | 1221 | |||
GenBank mRNA | AB030499.1 | - | 11 | 1156 | ||
AB030500.1 | - | 12 | 1210 | |||
AB030501.1 | - | 11 | 1156 |